Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mca-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268315.3 | NP_001255244.1 | 9 - 10 | 1203 | 63 | |
NM_001307544.3 | NP_001294473.1 | 3 - 4 | 260 | 63 | ||
NM_001383061.1 | 8 - 9 | 1100 | 63 | |||
NM_001383062.1 | 8 - 9 | 1102 | 63 | |||
NM_001383063.1 | 8 - 9 | 1102 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |