Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307526.4 | NP_001294455.1 | 1 - 2 | 113 | 57 | |
NM_067995.4 | NP_500396.2 | 1 - 2 | 117 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |