Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307366.3 | NP_001294295.1 | 5 - 6 | 989 | 98 | |
NM_068009.7 | NP_500410.2 | 5 - 6 | 1085 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |