Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zag-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068023.7 | NP_500424.3 | 2 - 3 | 709 | 67 | |
GenBank mRNA | AY224511.1 | - | 2 - 3 | 714 | 67 | |
AY289599.1 | - | 2 - 3 | 721 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |