Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068053.5 | NP_500454.2 | 11 - 12 | 3006 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |