Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028240.6 | NP_001023411.1 | 5 - 6 | 705 | 57 | |
NM_001028242.6 | NP_001023413.1 | 3 - 4 | 409 | 57 | ||
NM_001028243.3 | NP_001023414.1 | 3 - 4 | 574 | 57 | ||
NM_001364041.3 | NP_001350991.1 | 4 - 5 | 436 | 57 | ||
NM_001372787.3 | NP_001360026.1 | 4 - 5 | 649 | 57 | ||
NM_001380173.1 | NP_001368701.1 | 5 - 6 | 511 | 57 | ||
NM_001383072.1 | 5 - 6 | 674 | 57 | |||
NM_001383073.1 | 5 - 6 | 645 | 57 | |||
NM_068060.5 | NP_500461.1 | 4 - 5 | 630 | 57 | ||
NM_068061.6 | NP_500462.1 | 4 - 5 | 647 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |