Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372791.1 | NP_001360017.1 | 3 - 4 | 363 | 82 |
Gene Symbol: cyp-25A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047385.4 | NP_001040850.1 | 4 - 5 | 568 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |