Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068145.3 | NP_500546.2 | 8 - 9 | 1418 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |