Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068178.5 | NP_500579.3 | 2 - 3 | 382 | 79 | |
GenBank mRNA | EU729335.1 | - | 2 - 3 | 243 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |