Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047475.5 | NP_001040940.1 | 4 - 5 | 507 | 118 | |
NM_001047476.5 | NP_001040941.1 | 1 - 2 | 80 | 118 | ||
NM_001307453.3 | NP_001294382.1 | 4 - 5 | 476 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |