Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoe-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027938.6 | NP_001023109.1 | 5 - 6 | 2196 | 139 | |
NM_001027939.4 | NP_001023110.1 | 3 - 4 | 1957 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |