Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380207.2 | NP_001367921.1 | 2 - 3 | 378 | 92 | |
GenBank mRNA | AF502568.1 | - | 3 - 4 | 486 | 92 | |
AF519108.1 | - | 2 - 3 | 367 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |