Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380207.2 | NP_001367921.1 | 6 - 7 | 1061 | 134 | |
GenBank mRNA | AF502568.1 | - | 7 - 8 | 1169 | 134 | |
AF519108.1 | - | 6 - 7 | 1050 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |