Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306908.3 | NP_001293837.1 | 3 - 4 | 438 | 150 | |
NM_068407.5 | NP_500808.1 | 3 - 4 | 411 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |