Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ehn-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028444.3 | NP_001023615.1 | 4 - 5 | 516 | 122 | |
NM_001028445.1 | NP_001023616.1 | 2 - 3 | 357 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |