Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380231.1 | NP_001368360.1 | 4 - 5 | 1623 | 103 | |
NM_068414.6 | NP_500815.2 | 5 - 6 | 1928 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |