Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fem-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068423.6 | NP_500824.1 | 9 - 10 | 1263 | 111 | |
NM_068424.4 | NP_500825.1 | 10 - 11 | 1229 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |