Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkh-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027976.6 | NP_001023147.1 | 2 - 3 | 730 | 92 | |
NM_001027977.5 | NP_001023148.1 | 1 - 2 | 157 | 92 | ||
NM_001306882.4 | NP_001293811.1 | 2 - 3 | 715 | 92 | ||
NM_001306883.3 | NP_001293812.1 | 1 - 2 | 157 | 92 | ||
NM_001306884.4 | NP_001293813.1 | 2 - 3 | 715 | 92 | ||
NM_001306885.3 | NP_001293814.1 | 2 - 3 | 715 | 92 | ||
NM_001306886.3 | NP_001293815.1 | 1 - 2 | 157 | 92 | ||
NM_001306887.3 | NP_001293816.1 | 1 - 2 | 157 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |