Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkh-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027976.6 | NP_001023147.1 | 8 - 9 | 2132 | 70 | |
NM_001027977.5 | NP_001023148.1 | 7 - 8 | 1559 | 70 | ||
NM_001306791.3 | NP_001293720.1 | 4 - 5 | 668 | 70 | ||
NM_001306792.3 | NP_001293721.1 | 1 - 2 | 80 | 70 | ||
NM_001306794.3 | NP_001293723.1 | 4 - 5 | 668 | 70 | ||
NM_001306795.3 | NP_001293724.1 | 4 - 5 | 611 | 70 | ||
NM_001306882.4 | NP_001293811.1 | 8 - 9 | 2060 | 70 | ||
NM_001306883.3 | NP_001293812.1 | 7 - 8 | 1502 | 70 | ||
NM_001306884.4 | NP_001293813.1 | 8 - 9 | 2117 | 70 | ||
NM_001306885.3 | NP_001293814.1 | 8 - 9 | 2060 | 70 | ||
NM_001306886.3 | NP_001293815.1 | 7 - 8 | 1559 | 70 | ||
NM_001306887.3 | NP_001293816.1 | 7 - 8 | 1502 | 70 | ||
NM_001380234.1 | NP_001368377.1 | 4 - 5 | 611 | 70 | ||
NM_001380235.1 | NP_001368378.1 | 1 - 2 | 80 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |