Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068437.1 | NP_500838.1 | 1 - 2 | 276 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |