Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356923.3 | NP_001343801.1 | 1 - 2 | 271 | 85 | |
NM_068438.4 | NP_500839.2 | 2 - 3 | 706 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |