Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380249.1 | NP_001367345.1 | 6 - 7 | 2652 | 74 | |
NM_001380250.2 | NP_001367343.1 | 6 - 7 | 2671 | 74 | ||
NM_001380251.2 | NP_001367344.1 | 6 - 7 | 2668 | 74 | ||
NM_001380252.2 | NP_001368407.1 | 6 - 7 | 2657 | 74 | ||
NM_001380253.1 | NP_001368408.1 | 1 - 2 | 153 | 74 | ||
NM_001380254.1 | NP_001368409.1 | 1 - 2 | 153 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |