Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306874.3 | NP_001293803.1 | 1 - 2 | 179 | 84 | |
NM_001306875.3 | NP_001293804.1 | 1 - 2 | 179 | 84 | ||
NM_001306876.3 | NP_001293805.1 | 1 - 2 | 179 | 84 | ||
NM_001306877.4 | NP_001293806.1 | 1 - 2 | 179 | 84 | ||
NM_001306878.3 | NP_001293807.1 | 1 - 2 | 179 | 84 | ||
NM_001306879.3 | NP_001293808.1 | 1 - 2 | 179 | 84 | ||
NM_001306880.3 | NP_001293809.1 | 1 - 2 | 179 | 84 | ||
NM_068506.3 | NP_500907.2 | 1 - 2 | 179 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |