Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068547.2 | NP_500948.2 | 4 - 5 | 745 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |