Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028211.4 | NP_001023382.1 | 10 - 11 | 1395 | 125 | |
NR_131543.1 | 10 - 11 | 1404 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |