Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Y73B6BL.29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380266.1 | NP_001368403.1 | 6 - 7 | 1968 | 59 | |
NM_068561.6 | NP_500962.1 | 7 - 8 | 2147 | 59 |
Gene Symbol: sqd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380267.1 | NP_001368063.1 | 2 - 3 | 634 | 59 | |
NM_001380268.1 | NP_001368062.1 | 2 - 3 | 413 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |