Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mif-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068567.2 | NP_500968.1 | 2 - 3 | 192 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |