Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069769.5 | NP_502170.4 | 7 - 8 | 704 | 101 | |
NR_101961.1 | 7 - 8 | 652 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |