Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276863.2 | NP_001263792.1 | 4 - 5 | 746 | 113 | |
NM_069769.5 | NP_502170.4 | 13 - 14 | 1760 | 113 | ||
NR_101961.1 | 13 - 14 | 1708 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |