Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069772.8 | NP_502173.2 | 5 - 6 | 2200 | 69 | |
GenBank mRNA | AF416567.1 | - | 5 - 6 | 2222 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |