Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scl-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069827.3 | NP_502228.2 | 3 - 4 | 632 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |