Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lips-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083237.3 | NP_001076706.1 | 7 - 8 | 894 | 88 | |
NM_001083238.2 | NP_001076707.1 | 1 - 2 | 42 | 88 | ||
NM_001268659.2 | NP_001255588.1 | 8 - 9 | 1033 | 88 | ||
NM_001268660.3 | NP_001255589.1 | 8 - 9 | 939 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |