Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028457.5 | NP_001023628.1 | 1 - 2 | 89 | 76 | |
GenBank mRNA | AF497834.1 | - | 1 - 2 | 111 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |