Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: nhx-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028456.4 | NP_001023627.1 | 2 - 3 | 494 | 75 | |
NM_001028457.5 | NP_001023628.1 | 2 - 3 | 172 | 75 | ||
GenBank mRNA | AF497834.1 | - | 2 - 3 | 194 | 75 | |
AF497835.1 | - | 2 - 3 | 446 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |