Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268695.2 | NP_001255624.1 | 4 - 5 | 585 | 67 | |
NM_001268696.3 | NP_001255625.1 | 2 - 3 | 264 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |