Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069911.5 | NP_502312.2 | 20 - 21 | 4202 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |