Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372935.2 | NP_001360368.1 | 3 | 622 | 131 | |
NM_069944.3 | NP_502345.1 | 2 - 3 | 547 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |