Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069972.5 | NP_502373.3 | 7 - 8 | 1152 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |