Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356933.3 | NP_001343588.1 | 5 | 1164 | 150 | |
NM_001356934.1 | NP_001343704.1 | 1 - 2 | 68 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |