Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070011.5 | NP_502412.1 | 5 - 6 | 2019 | 104 | |
NR_131627.1 | 6 - 7 | 1914 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |