Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ark-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268724.2 | NP_001255653.1 | 1 - 2 | 126 | 87 | |
NM_001268725.3 | NP_001255654.1 | 1 - 2 | 126 | 87 | ||
GenBank mRNA | AJ271057.1 | - | 1 - 2 | 126 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |