Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 14 - 15 | 1963 | 76 | ||
NM_070018.6 | NP_502419.2 | 9 - 10 | 1231 | 76 | ||
GenBank mRNA | AB055112.1 | - | 11 - 12 | 1628 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |