Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268734.3 | NP_001255663.1 | 15 - 16 | 3001 | 94 | |
NM_001268735.4 | NP_001255664.1 | 14 - 15 | 2980 | 94 | ||
NM_001268736.3 | NP_001255665.1 | 13 - 14 | 2944 | 94 | ||
NM_001268737.4 | NP_001255666.1 | 15 - 16 | 3035 | 94 | ||
NM_001268738.3 | NP_001255667.1 | 14 - 15 | 3004 | 94 | ||
NM_001268739.3 | NP_001255668.1 | 6 - 7 | 1636 | 94 | ||
NM_001268740.3 | NP_001255669.1 | 6 - 7 | 1636 | 94 | ||
NM_001306972.3 | NP_001293901.1 | 17 - 18 | 3049 | 94 | ||
NM_001306973.3 | NP_001293902.1 | 16 - 17 | 3037 | 94 | ||
NM_001306974.3 | NP_001293903.1 | 16 - 17 | 3028 | 94 | ||
NM_001306975.3 | NP_001293904.1 | 15 - 16 | 3016 | 94 | ||
NM_001306976.3 | NP_001293905.1 | 15 - 16 | 2992 | 94 | ||
NM_001306977.3 | NP_001293906.1 | 14 - 15 | 2980 | 94 | ||
NM_001306978.3 | NP_001293907.1 | 16 - 17 | 3013 | 94 | ||
NM_001306979.3 | NP_001293908.1 | 15 - 16 | 2992 | 94 | ||
NM_001306980.3 | NP_001293909.1 | 14 - 15 | 2956 | 94 | ||
NM_001306981.3 | NP_001293910.1 | 7 - 8 | 1648 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |