Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268734.3 | NP_001255663.1 | 16 - 17 | 3142 | 90 | |
NM_001268735.4 | NP_001255664.1 | 15 - 16 | 3121 | 90 | ||
NM_001268736.3 | NP_001255665.1 | 14 - 15 | 3085 | 90 | ||
NM_001268737.4 | NP_001255666.1 | 16 - 17 | 3176 | 90 | ||
NM_001268738.3 | NP_001255667.1 | 15 - 16 | 3145 | 90 | ||
NM_001268739.3 | NP_001255668.1 | 7 - 8 | 1777 | 90 | ||
NM_001268740.3 | NP_001255669.1 | 7 - 8 | 1777 | 90 | ||
NM_001306972.3 | NP_001293901.1 | 18 - 19 | 3190 | 90 | ||
NM_001306973.3 | NP_001293902.1 | 17 - 18 | 3178 | 90 | ||
NM_001306974.3 | NP_001293903.1 | 17 - 18 | 3169 | 90 | ||
NM_001306975.3 | NP_001293904.1 | 16 - 17 | 3157 | 90 | ||
NM_001306976.3 | NP_001293905.1 | 16 - 17 | 3133 | 90 | ||
NM_001306977.3 | NP_001293906.1 | 15 - 16 | 3121 | 90 | ||
NM_001306978.3 | NP_001293907.1 | 17 - 18 | 3154 | 90 | ||
NM_001306979.3 | NP_001293908.1 | 16 - 17 | 3133 | 90 | ||
NM_001306980.3 | NP_001293909.1 | 15 - 16 | 3097 | 90 | ||
NM_001306981.3 | NP_001293910.1 | 8 - 9 | 1789 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |