Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028036.4 | NP_001023207.1 | 7 - 8 | 1117 | 78 | |
NM_001028037.4 | NP_001023208.1 | 7 - 8 | 1111 | 78 | ||
NM_001047485.5 | NP_001040950.1 | 1 - 2 | 130 | 78 | ||
NM_001047486.4 | NP_001040951.1 | 2 - 3 | 423 | 78 | ||
NM_001268766.4 | NP_001255695.1 | 2 - 3 | 312 | 78 | ||
NM_001306985.3 | NP_001293914.1 | 2 - 3 | 229 | 78 | ||
NM_001306986.3 | NP_001293915.1 | 1 - 2 | 130 | 78 | ||
NM_001306987.4 | NP_001293916.1 | 8 - 9 | 1060 | 78 | ||
NM_001306988.3 | NP_001293917.1 | 2 - 3 | 265 | 78 | ||
NM_001380497.1 | NP_001368118.1 | 8 - 9 | 1060 | 78 | ||
NM_001380498.1 | NP_001368149.1 | 2 - 3 | 265 | 78 | ||
NM_001380499.1 | NP_001368473.1 | 2 - 3 | 229 | 78 | ||
NM_001380500.1 | NP_001366773.1 | 2 - 3 | 229 | 78 | ||
NM_001380501.1 | NP_001368474.1 | 1 - 2 | 130 | 78 | ||
GenBank mRNA | AY090019.1 | - | 7 - 8 | 1134 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |