Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028036.4 | NP_001023207.1 | 8 - 9 | 1426 | 86 | |
NM_001028037.4 | NP_001023208.1 | 8 - 9 | 1420 | 86 | ||
NM_001047485.5 | NP_001040950.1 | 2 - 3 | 439 | 86 | ||
NM_001047486.4 | NP_001040951.1 | 3 - 4 | 732 | 86 | ||
NM_001268766.4 | NP_001255695.1 | 3 - 4 | 621 | 86 | ||
NM_001306985.3 | NP_001293914.1 | 3 - 4 | 538 | 86 | ||
NM_001306986.3 | NP_001293915.1 | 2 - 3 | 439 | 86 | ||
NM_001306987.4 | NP_001293916.1 | 9 - 10 | 1369 | 86 | ||
NM_001306988.3 | NP_001293917.1 | 3 - 4 | 574 | 86 | ||
NM_001380497.1 | NP_001368118.1 | 9 - 10 | 1369 | 86 | ||
NM_001380498.1 | NP_001368149.1 | 3 - 4 | 574 | 86 | ||
NM_001380499.1 | NP_001368473.1 | 3 - 4 | 538 | 86 | ||
NM_001380500.1 | NP_001366773.1 | 3 - 4 | 538 | 86 | ||
NM_001380501.1 | NP_001368474.1 | 2 - 3 | 439 | 86 | ||
GenBank mRNA | AY090019.1 | - | 8 - 9 | 1443 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |