Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026105.4 | NP_001021276.1 | 3 - 4 | 605 | 63 | |
NM_001026106.2 | NP_001021277.1 | 3 - 4 | 492 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |