Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lev-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268775.4 | NP_001255704.1 | 1 - 2 | 248 | 63 | |
NM_001268776.2 | NP_001255705.1 | 1 - 2 | 385 | 63 | ||
GenBank mRNA | X98601.1 | - | 298 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |