Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lev-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268775.4 | NP_001255704.1 | 2 - 3 | 569 | 118 | |
NM_001268776.2 | NP_001255705.1 | 2 - 3 | 706 | 118 | ||
GenBank mRNA | X98601.1 | - | 619 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |