Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lev-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268775.4 | NP_001255704.1 | 3 - 4 | 722 | 96 | |
NM_001268776.2 | NP_001255705.1 | 3 - 4 | 859 | 96 | ||
GenBank mRNA | X98601.1 | - | 772 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |